Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006695.1 | NP_001006696.1 | 7 - 8 | 795 | 89 | |
GenBank mRNA | BC075400.1 | - | 7 - 8 | 795 | 89 | |
CR761026.2 | - | 7 - 8 | 769 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |