Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006817.1 | NP_001006818.1 | 4 - 5 | 753 | 91 | |
GenBank mRNA | BC076712.1 | - | 4 - 5 | 753 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |