Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006830.1 | NP_001006831.1 | 1 - 2 | 326 | 70 | |
GenBank mRNA | BC076899.1 | - | 1 - 2 | 326 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |