Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006912.1 | NP_001006913.1 | 2 - 3 | 164 | 64 | |
XM_012970316.1 | XP_012825770.1 | 2 - 3 | 182 | 64 | ||
XM_018097088.1 | XP_017952577.1 | 1 - 2 | 88 | 64 | ||
GenBank mRNA | BC075391.1 | - | 2 - 3 | 164 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |