Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007902.1 | NP_001007903.1 | 3 - 4 | 621 | 82 | |
GenBank mRNA | BC080339.1 | - | 3 - 4 | 621 | 82 | |
CR848299.2 | - | 3 - 4 | 610 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |