Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008067.1 | NP_001008068.1 | 2 - 3 | 373 | 76 | |
GenBank mRNA | BC080959.1 | - | 2 - 3 | 373 | 76 | |
CR855837.1 | - | 2 - 3 | 304 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |