Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cidec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008101.1 | NP_001008102.1 | 6 - 7 | 719 | 68 | |
XM_012960954.2 | XP_012816408.1 | 5 - 6 | 1263 | 68 | ||
XM_012960955.2 | XP_012816409.1 | 7 - 8 | 1018 | 68 | ||
XM_012960956.2 | XP_012816410.1 | 5 - 6 | 707 | 68 | ||
XM_012960957.2 | XP_012816411.1 | 5 - 6 | 692 | 68 | ||
XM_018093023.1 | XP_017948512.1 | 6 - 7 | 916 | 68 | ||
GenBank mRNA | BC081304.1 | - | 6 - 7 | 719 | 68 | |
CR761715.2 | - | 6 - 7 | 727 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |