Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011151.1 | NP_001011151.1 | 4 - 5 | 572 | 101 | |
GenBank mRNA | BC084901.1 | - | 4 - 5 | 572 | 101 | |
CR760404.1 | - | 4 - 5 | 458 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |