Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011194.1 | NP_001011194.1 | 1 - 2 | 288 | 70 | |
GenBank mRNA | BC087608.1 | - | 1 - 2 | 288 | 70 | |
CR760467.2 | - | 1 - 2 | 263 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |