Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203588.1 | NP_988919.1 | 7 - 8 | 817 | 99 | |
GenBank mRNA | BC061621.1 | - | 7 - 8 | 817 | 99 | |
BC080151.1 | - | 7 - 8 | 828 | 99 | ||
CR762274.2 | - | 7 - 8 | 810 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |