Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cxorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204012.2 | NP_989343.1 | 1 - 2 | 158 | 69 | |
XM_018096190.1 | XP_017951679.1 | 1 - 2 | 214 | 69 | ||
GenBank mRNA | BC064195.1 | - | 1 - 2 | 139 | 69 | |
BC084906.1 | - | 1 - 2 | 158 | 69 | ||
CR760470.2 | - | 1 - 2 | 139 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |