Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011415.1 | NP_001011415.1 | 3 - 4 | 305 | 74 | |
XM_012961302.2 | XP_012816756.1 | 4 - 5 | 799 | 74 | ||
GenBank mRNA | BC087984.1 | - | 3 - 4 | 305 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |