Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015722.1 | NP_001015722.1 | 1074 | 86 | ||
GenBank mRNA | BC089654.1 | - | 1074 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis |