Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st3gal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016104.3 | NP_001016104.2 | 4 - 5 | 625 | 84 | |
XM_012960776.2 | XP_012816230.1 | 3 - 4 | 2015 | 84 | ||
XM_012960777.2 | XP_012816231.1 | 4 - 5 | 2065 | 84 | ||
GenBank mRNA | BC121863.1 | - | 4 - 5 | 625 | 84 | |
CR762128.2 | - | 4 - 5 | 661 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |