Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016109.2 | NP_001016109.1 | 3 - 4 | 964 | 75 | |
GenBank mRNA | BC170592.1 | - | 3 - 4 | 822 | 75 | |
BC170596.1 | - | 3 - 4 | 822 | 75 | ||
CR762116.2 | - | 3 - 4 | 964 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |