Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pitpnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016639.2 | NP_001016639.1 | 5 - 6 | 466 | 106 | |
XM_012966615.2 | XP_012822069.1 | 5 - 6 | 602 | 106 | ||
XM_018095393.1 | XP_017950882.1 | 5 - 6 | 605 | 106 | ||
GenBank mRNA | BC118687.1 | - | 5 - 6 | 406 | 106 | |
BC171116.1 | - | 5 - 6 | 393 | 106 | ||
CR760415.2 | - | 5 - 6 | 466 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |