Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016749.1 | NP_001016749.1 | 6 - 7 | 597 | 85 | |
GenBank mRNA | BC158150.1 | - | 6 - 7 | 741 | 85 | |
CR855811.1 | - | 6 - 7 | 597 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |