Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017066.2 | NP_001017066.1 | 5 - 6 | 453 | 97 | |
XM_012957024.2 | XP_012812478.1 | 5 - 6 | 499 | 97 | ||
XM_018091272.1 | XP_017946761.1 | 5 - 6 | 499 | 97 | ||
GenBank mRNA | CR761601.2 | - | 5 - 6 | 453 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |