Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017188.2 | NP_001017188.1 | 4 - 5 | 687 | 98 | |
XM_018092112.1 | XP_017947601.1 | 4 - 5 | 862 | 98 | ||
GenBank mRNA | CR760622.2 | - | 4 - 5 | 687 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |