Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017196.2 | NP_001017196.1 | 3 - 4 | 393 | 105 | |
XM_012963420.1 | XP_012818874.1 | 3 - 4 | 563 | 105 | ||
GenBank mRNA | CR760545.2 | - | 3 - 4 | 393 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |