Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017196.2 | NP_001017196.1 | 7 - 8 | 878 | 100 | |
GenBank mRNA | CR760545.2 | - | 7 - 8 | 878 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |