Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017225.2 | NP_001017225.1 | 1 - 2 | 192 | 61 | |
XM_012957048.2 | XP_012812502.1 | 2 - 3 | 253 | 61 | ||
XM_018091279.1 | XP_017946768.1 | 2 - 3 | 316 | 61 | ||
GenBank mRNA | BC159011.1 | - | 1 - 2 | 148 | 61 | |
BC161583.1 | - | 1 - 2 | 148 | 61 | ||
CR760297.2 | - | 1 - 2 | 192 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |