Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017309.2 | NP_001017309.1 | 3 - 4 | 587 | 70 | |
GenBank mRNA | CR855514.2 | - | 3 - 4 | 587 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |