Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030330.1 | NP_001025501.1 | 5 - 6 | 818 | 83 | |
GenBank mRNA | BC080175.1 | - | 5 - 6 | 824 | 83 | |
BC161475.1 | - | 5 - 6 | 811 | 83 | ||
CR760257.1 | - | 5 - 6 | 818 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |