Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hao2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030453.1 | NP_001025624.1 | 6 - 7 | 1006 | 67 | |
XM_012957078.2 | XP_012812532.1 | 7 - 8 | 1205 | 67 | ||
XM_012957079.2 | XP_012812533.1 | 7 - 8 | 1202 | 67 | ||
XM_012957080.2 | XP_012812534.1 | 6 - 7 | 1076 | 67 | ||
XM_012957081.2 | XP_012812535.1 | 7 - 8 | 1274 | 67 | ||
XM_012957082.2 | XP_012812536.1 | 6 - 7 | 1497 | 67 | ||
XM_012957084.1 | XP_012812538.1 | 6 - 7 | 1098 | 67 | ||
XM_012957085.2 | XP_012812539.1 | 6 - 7 | 1494 | 67 | ||
XM_012957086.2 | XP_012812540.1 | 6 - 7 | 1538 | 67 | ||
XM_012957087.2 | XP_012812541.1 | 6 - 7 | 1150 | 67 | ||
GenBank mRNA | BC091092.1 | - | 6 - 7 | 1006 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |