Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032327.1 | NP_001027498.1 | 21 - 22 | 3160 | 89 | |
GenBank mRNA | BC096519.1 | - | 21 - 22 | 3160 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |