Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011074.2 | NP_001011074.1 | 6 - 7 | 1058 | 103 | |
XM_012956842.2 | XP_012812296.1 | 6 - 7 | 1462 | 103 | ||
GenBank mRNA | BC084459.1 | - | 6 - 7 | 1001 | 103 | |
CR848150.2 | - | 6 - 7 | 1058 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |