Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: entpd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015918.2 | NP_001015918.1 | 11 - 12 | 1580 | 104 | |
XM_012966551.2 | XP_012822005.1 | 11 - 12 | 1617 | 104 | ||
GenBank mRNA | BC170622.1 | - | 10 - 11 | 1543 | 104 | |
CR926412.2 | - | 11 - 12 | 1580 | 104 | ||
DQ118412.1 | - | 9 - 10 | 1428 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |