Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015961.2 | NP_001015961.1 | 5 - 6 | 1100 | 81 | |
GenBank mRNA | BC122010.1 | - | 5 - 6 | 972 | 81 | |
BC161506.1 | - | 5 - 6 | 955 | 81 | ||
CR848436.2 | - | 5 - 6 | 1100 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |