Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016111.2 | NP_001016111.1 | 1 - 2 | 245 | 78 | |
GenBank mRNA | BC155533.1 | - | 1 - 2 | 280 | 78 | |
CR762112.2 | - | 1 - 2 | 245 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |