Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016157.2 | NP_001016157.1 | 2 - 3 | 190 | 83 | |
GenBank mRNA | CR761936.2 | - | 2 - 3 | 190 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |