Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b4galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016385.2 | NP_001016385.1 | 4 - 5 | 843 | 61 | |
GenBank mRNA | BC170760.1 | - | 4 - 5 | 629 | 61 | |
BC170764.1 | - | 4 - 5 | 629 | 61 | ||
CR761207.2 | - | 4 - 5 | 843 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |