Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016431.2 | NP_001016431.1 | 3 - 4 | 383 | 80 | |
GenBank mRNA | BC135174.2 | - | 3 - 4 | 316 | 80 | |
CR761051.2 | - | 3 - 4 | 383 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |