Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016632.2 | NP_001016632.1 | 4 - 5 | 557 | 82 | |
GenBank mRNA | BC168415.1 | - | 4 - 5 | 503 | 82 | |
BC170617.1 | - | 4 - 5 | 382 | 82 | ||
BC170619.1 | - | 4 - 5 | 380 | 82 | ||
CR760445.2 | - | 4 - 5 | 557 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |