Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037272.1 | NP_001032349.1 | 2 - 3 | 248 | 97 | |
GenBank mRNA | BC077668.1 | - | 2 - 3 | 253 | 97 | |
BC088002.1 | - | 2 - 3 | 248 | 97 | ||
CR848186.2 | - | 2 - 3 | 197 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |