Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204041.2 | NP_989372.1 | 1 - 2 | 221 | 73 | |
GenBank mRNA | BC064191.1 | - | 1 - 2 | 218 | 73 | |
CR855594.2 | - | 1 - 2 | 221 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |