Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204041.2 | NP_989372.1 | 5 - 6 | 782 | 102 | |
GenBank mRNA | BC064191.1 | - | 5 - 6 | 779 | 102 | |
CR855594.2 | - | 5 - 6 | 782 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |