Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tuba3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015934.4 | NP_001015934.1 | 1258 | 93 | ||
GenBank mRNA | BC080160.1 | - | 3 - 4 | 508 | 93 | |
BC135250.1 | - | 3 - 4 | 490 | 93 | ||
CR926373.2 | - | 3 - 4 | 478 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |