Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045569.1 | NP_001039034.1 | 3 - 4 | 692 | 80 | |
XM_012961256.1 | XP_012816710.1 | 3 - 4 | 772 | 80 | ||
GenBank mRNA | CR762182.2 | - | 3 - 4 | 692 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Xenopus tropicalis Probe spans exons |