Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045663.1 | NP_001039128.1 | 7 - 8 | 1453 | 83 | |
GenBank mRNA | BC160567.1 | - | 6 - 7 | 1442 | 83 | |
CR942486.1 | - | 7 - 8 | 1453 | 83 |
Gene Symbol: LOC100327245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171528.1 | NP_001164999.1 | 6 - 7 | 1442 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |