Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ogfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045746.1 | NP_001039211.1 | 5 - 6 | 527 | 68 | |
XM_012964506.2 | XP_012819960.1 | 5 - 6 | 885 | 68 | ||
XM_012964510.2 | XP_012819964.1 | 4 - 5 | 426 | 68 | ||
XM_012964515.2 | XP_012819969.1 | 6 - 7 | 719 | 68 | ||
XM_018094113.1 | XP_017949602.1 | 5 - 6 | 492 | 68 | ||
XM_018094117.1 | XP_017949606.1 | 5 - 6 | 414 | 68 | ||
XM_018094120.1 | XP_017949609.1 | 4 - 5 | 381 | 68 | ||
GenBank mRNA | BC157323.1 | - | 6 - 7 | 724 | 68 | |
CR942500.2 | - | 5 - 6 | 527 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |