Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045770.1 | NP_001039235.1 | 7 - 8 | 677 | 91 | |
GenBank mRNA | BC157333.1 | - | 7 - 8 | 711 | 91 | |
CR942713.1 | - | 7 - 8 | 677 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |