Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078721.1 | NP_001072189.1 | 5 - 6 | 711 | 86 | |
XM_012953410.2 | XP_012808864.1 | 4 - 5 | 726 | 86 | ||
GenBank mRNA | BC118723.1 | - | 5 - 6 | 711 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |