Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfrsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079136.1 | NP_001072604.1 | 3 - 4 | 1339 | 66 | |
XM_012970559.2 | XP_012826013.1 | 3 - 4 | 1365 | 66 | ||
GenBank mRNA | BC122979.1 | - | 3 - 4 | 1339 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |