Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079143.1 | NP_001072611.1 | 6 - 7 | 1386 | 108 | |
XM_012965635.2 | XP_012821089.1 | 6 - 7 | 1380 | 108 | ||
GenBank mRNA | BC122993.1 | - | 6 - 7 | 1386 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |