Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079245.1 | NP_001072713.1 | 674 | 62 | ||
GenBank mRNA | BC125667.1 | - | 674 | 62 | ||
CT030525.1 | - | 3 - 4 | 554 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |