Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079262.2 | NP_001072730.2 | 7 - 8 | 1523 | 108 | |
GenBank mRNA | BC125705.1 | - | 7 - 8 | 1523 | 108 | |
BC135434.1 | - | 7 - 8 | 1446 | 108 | ||
CT025323.2 | - | 7 - 8 | 1430 | 108 | ||
CX740928.1 | - | 699 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |