Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079446.1 | NP_001072914.1 | 5 - 6 | 352 | 78 | |
XM_012959300.2 | XP_012814754.1 | 5 - 6 | 479 | 78 | ||
XM_018092187.1 | XP_017947676.1 | 4 - 5 | 354 | 78 | ||
GenBank mRNA | BC118825.1 | - | 5 - 6 | 352 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |