Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017049.3 | NP_001017049.1 | 4 - 5 | 554 | 98 | |
GenBank mRNA | BC135615.1 | - | 4 - 5 | 554 | 98 | |
CR761816.2 | - | 4 - 5 | 467 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |