Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079092.2 | NP_001072560.2 | 467 | 90 | ||
GenBank mRNA | DR870839.1 | - | 1 - 2 | 98 | 90 | |
DR872131.1 | - | 3 - 4 | 467 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |